Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BMPR1A/BMPR1B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5143395
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
Encapsidates the viral DNA into characteristic twinned ('geminate') particles Binds the genomic viral ssDNA and shuttles it into and out of the cell nucleus The CP of bipartite geminiviruses is not required for cell-to-cell or systemic movement.
Specifications
| BMPR1A/BMPR1B | |
| Polyclonal | |
| Unconjugated | |
| alk6; alk6b; alk6tr; bmpr1b; bmpr1ba; bmpr1bb; BMPR-IB; bone morphogenetic protein receptor type 1b; bone morphogenetic protein receptor type 1ba; bone morphogenetic protein receptor type 1bb; bone morphogenetic protein receptor type IB,b; bone morphogenetic protein receptor type-1B; BR1b; MGC172219; MGC92220; zALK-6; zgc:172219; zgc:92220 | |
| The immunogen used to generate this antibody corresponds to zebrafish Bmpr1ba+b. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| 0.1 mg/mL | |
| PBS with 20% glycerol and 0.025% Proclin 300; pH 7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| Zebrafish | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction