Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BMPR1A Monoclonal Antibody (4B7B2)
GREENER_CHOICE

Catalog No. PIMA517036
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517036 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA517036 Supplier Invitrogen™ Supplier No. MA517036
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-17036 targets BMPR1A in indirect ELISA, FACS, IHC and WB applications and shows reactivity with Human samples. The MA5-17036 immunogen is purified recombinant fragment of human BMPR1A (amino acids: 179-378 ) expressed in E. Coli. MA5-17036 detects BMPR1A which has a predicted molecular weight of approximately 60kDa.

BMPR1A is a Type I member of the TGF beta receptor superfamily of transmembrane ser/thr kinases which phosphorylates intracellular SMADs in response to bone morphogenic proteins. These receptors are also closely related to the activin receptors, ACVR1 and ACVR2. The ligands of these receptors are members of the TGF-beta superfamily. TGF-betas and activins transduce their signals through the formation of heteromeric complexes with 2 different types of serine (threonine) kinase receptors: type I receptors of about 50-55 kD and type II receptors of about 70-80 kD. Type II receptors bind ligands in the absence of type I receptors, but they require their respective type I receptors for signaling, whereas type I receptors require their respective type II receptors for ligand binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BMPR1A
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 4B7B2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene BMPR1A
Gene Accession No. P36894
Gene Alias 10q23del; 1110037I22Rik; activin A receptor, type II-like kinase 3; activin receptor-like kinase 3; ACVRLK3; ALK3; ALK-3; AU045487; BMP type-1A receptor; BMP-2/BMP-4 receptor; Bmpr; Bmpr1a; BMPR-1A; BMPRIA; BMPR-IA; Bone morphogenetic protein 4 receptor; bone morphogenetic protein receptor type 1A; bone morphogenetic protein receptor type IA; bone morphogenetic protein receptor type-1A; bone morphogenetic protein receptor, type 1A; bone morphogenetic protein receptor, type IA; CD292; Serine/threonine-protein kinase receptor R5; SKR5
Gene Symbols BMPR1A
Host Species Mouse
Immunogen Purified recombinant fragment of human BMPR1A (amino acids: 179-378 ) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 657
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.