Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMPR1B Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987395
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7395 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7395 Supplier Bioss Supplier No. BS6639R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The bone morphogenetic protein (BMP) receptors are a family of transmembrane serine/threonine kinases that include the type I receptors BMPR1A and BMPR1B and the type II receptor BMPR2. These receptors are also closely related to the activin receptors, ACVR1 and ACVR2. The ligands of these receptors are members of the TGF-beta superfamily. TGF-betas and activins transduce their signals through the formation of heteromeric complexes with 2 different types of serine (threonine) kinase receptors: type I receptors of about 50-55 kD and type II receptors of about 70-80 kD. Type II receptors bind ligands in the absence of type I receptors, but they require their respective type I receptors for signaling, whereas type I receptors require their respective type II receptors for ligand binding.

Specifications

Antigen BMPR1B
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene BMPR1B
Gene Accession No. O00238
Gene Alias Activin receptor-like kinase 6; Acvrlk6; AI385617; Alk6; ALK-6; AMDD; AV355320; BDA1D; BDA2; BMP type-1B receptor; BMP15; BMPR1B; BMPR-1B; BMPRIB; BMPR-IB; bone morphogenetic protein binding serine/threonine kinase receptor; bone morphogenetic protein receptor IB; bone morphogenetic protein receptor type 1B; bone morphogenetic protein receptor type IB; Bone morphogenetic protein receptor type-1B; bone morphogenetic protein receptor, type 1B; bone morphogenetic protein receptor, type IB; BOS_6477; CDw293; CFK-43a; serine/threonine receptor kinase; Serine/threonine-protein kinase receptor R6; SKR6
Gene Symbols BMPR1B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human BMPR1B.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 658
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.