Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BMS 265246, >97%
GREENER_CHOICESDP

Catalog No. 565450 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-545-0 50 mg
56-541-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 56-545-0 Supplier Tocris Bioscience™ Supplier No. 5654/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent cdk1/2 inhibitor

Potent cyclin-dependent kinase 1 (cdk1) and cdk 2 inhibitor (IC50 values are 5.6 - 71 and 6 - 14 nM, respectively). Also inhibits cdk4 and cdk5 in the sub micromolar range and cdk7 and cdk9 in the low micromolar range. Inhibits proliferation of HCT-116 colon cancer cells in vitro.

Chemical Identifiers

CAS 582315-72-8
Molecular Formula C18H17F2N3O2

Specifications

Quantity 50 mg
Solubility Soluble to 100mM in water
Formula Weight 345.34
Purity >97%
Molecular Weight (g/mol) 345.35
Chemical Name or Material (4-Butoxy-1H-pyrazolo[3,4-b]pyridin-5-yl) (2,6-difluoro-4-methylphenyl)methanone
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.