Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BMS 303141
GREENER_CHOICESDP

Catalog No. 460950 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-095-0 50 mg
46-091-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 46-095-0 Supplier Tocris Bioscience™ Supplier No. 4609/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

ATP citrate lyase inhibitor; orally bioavailable

ATP citrate lyase (ACL) inhibitor (IC50 = 0.13 μM for human recombinant ACL); blocks lipid synthesis (IC50 = 8 μM in HepG2 cells). Displays no cytotoxicity up to a concentration of 50 μM. Lowers plasma glucose and triglycerides in a mouse model of hyperlipidemia. Orally bioavailable.

Chemical Identifiers

CAS 943962-47-8
Molecular Formula C19H15Cl2NO4S

Specifications

Quantity 50 mg
Formula Weight 424.3
Purity 0.98
Molecular Weight (g/mol) 424.29
Chemical Name or Material BMS 303141
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.