Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BMS 345541
GSA_VAGREENER_CHOICESDP

Catalog No. 480650 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-065-0 50 mg
48-061-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 48-065-0 Supplier Tocris Bioscience™ Supplier No. 4806/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective allosteric inhibitor of IKK; anti-inflammatory

BMS 345541 is a selective allosteric inhibitor of IKK (IC50 values are 0.3 and 4.0 μM for IKKβ and IKKα respectively). Exhibits no effect against a panel of 15 other kinases. Attenuates LPS-induced cytokine production in vitro and blocks NFκB dependent transcription in mice. Also suppresses joint destruction in a mouse model of arthritis.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Quantity 50 mg
Product Type BMS 345541
Purity >99%
Molecular Weight (g/mol) 291.78
Inhibitors BMS 345541

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.