Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ BMS 536924
GREENER_CHOICESDP

Catalog No. 477410 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-741-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 47-741-0 Supplier R&D Systems™ Supplier No. 4774/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Dual IR/IGF1R inhibitor

Dual inhibitor of the insulin receptor (IR) and insulin-like growth factor-1 receptor (IGF1R) (IC50 values are 73 and 100 nM respectively). Inhibits receptor autophosphorylation and downstream MEK1/2 and Akt signaling. Induces G1 arrest and apoptosis in ML-1 cells; also inhibits cell proliferation in multiple tumor types. Reverses EMT through the attenuation of Snail mRNA expression in MCF10A cell over expressing IGF1R.

Chemical Identifiers

CAS 468740-43-4
Molecular Formula C25H26ClN5O3

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 479.96
Purity >98%
Molecular Weight (g/mol) 479.965
Chemical Name or Material BMS 536924
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.