Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BMS1/BMS1L Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | BMS1/BMS1L |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-9566
![]() |
Bethyl Laboratories, Inc
A305110AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
BMS1 (Ribosome biogenesis protein BMS1 homolog) is a 1,282 amino acid protein encoded by the human gene BMS1. BMS1 is a nuclear protein that belongs to the BMS1/TSR1 family (BMS1 subfamily). BMS1 is believed to act as a molecular switch during maturation of the 40S ribosomal subunit in the nucleolus. The 40S ribosomal subunit is an important member of the 80S ribosome complex, which also includes initiator tRNA and a 60S ribosomal subunit. The 80S ribosome is assembled by eukaryotic initiation factors (eIFs) at the initiation codon of mRNA in order to begin translation initiation. The joining of these ribosomal subunits requires eIF5B.Specifications
BMS1/BMS1L | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Ribosome biogenesis protein BMS1 homolog, Ribosome assembly protein BMS1 homolog, BMS1, BMS1L, KIAA0187 | |
BMS1 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q14692 | |
9790 | |
Between 1232 and 1282 | |
Primary | |
4° C | |
BMS1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title