Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BMS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560230
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560230 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560230 Supplier Invitrogen™ Supplier No. PA560230
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ELGGLFRVNQ PDRECKHKAD SLDCSRFLVE APHDWDLEEV MNSIRDCFVT GKWEDDKDAA KVLAEDEELY GDFEDLETGD VHKGKSGPNT QNEDI Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.

BMS1 (Ribosome biogenesis protein BMS1 homolog) is a 1,282 amino acid protein encoded by the human gene BMS1. BMS1 is a nuclear protein that belongs to the BMS1/TSR1 family (BMS1 subfamily). BMS1 is believed to act as a molecular switch during maturation of the 40S ribosomal subunit in the nucleolus. The 40S ribosomal subunit is an important member of the 80S ribosome complex, which also includes initiator tRNA and a 60S ribosomal subunit. The 80S ribosome is assembled by eukaryotic initiation factors (eIFs) at the initiation codon of mRNA in order to begin translation initiation. The joining of these ribosomal subunits requires eIF5B.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BMS1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Bms1
Gene Accession No. Q14692
Gene Alias AA408648; ACC; AU020092; AU043373; BB007109; BC030906; BMS1; BMS1 homolog, ribosome assembly protein; BMS1 homolog, ribosome assembly protein (yeast); BMS1 ribosome biogenesis factor; BMS1, ribosome biogenesis factor; Bms1l; BMS1-like, ribosome assembly protein; KIAA0187; mKIAA0187; ribosome assembly protein BMS1 homolog; Ribosome biogenesis protein BMS1 homolog
Gene Symbols Bms1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BMS1. Recombinant protein control fragment (Product #RP-102935).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9790
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.