Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BMX Antibody (1C6), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP147489
Description
BMX Monoclonal specifically detects BMX in Human samples. It is validated for Western Blot, ELISA.Specifications
BMX | |
Monoclonal | |
Unconjugated | |
BMX non-receptor tyrosine kinase, Bone marrow tyrosine kinase gene in chromosome X protein, EC 2.7.10, EC 2.7.10.2, Epithelial and endothelial tyrosine kinase, Etk/Bmx cytosolic tyrosine kinase, ETKcytoplasmic tyrosine-protein kinase BMX, NTK38, NTK38 tyrosine kinase, PSCTK2, PSCTK3 | |
Mouse | |
78 kDa | |
0.1 mL | |
Protein Kinase | |
660 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, ELISA | |
1C6 | |
Western Blot 1:500-1:2000, ELISA 1:10000 | |
BMX | |
Purified recombinant fragment of human BMX expressed in E. Coli. | |
Unpurified | |
RUO | |
Primary | |
Human | |
Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction