Learn More
Promega BMX Kinase Enzyme System, 10microg
Catalog No. PRV4512 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4512 10 μg
1 options
Catalog No. PRV4512 Supplier Promega Supplier No. V4512
Recombinant full-length human BMX was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. The BMX gene encodes a non-receptor tyrosine kinase, which may play a role in the growth and differentiation of hematopoietic cells.
  • Easily Screen and Profile BMX Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C