Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human BMX, His Tag Recombinant Protein

Catalog No. PV3371
Encompass_Preferred
Click to view available options
Quantity:
10 μg

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length, Recombinant, Cytoplasmic

Bone marrow tyrosine kinase (BMX) is a non-receptor tyrosine kinase that belongs to the SRC-related TEC subfamily of tyrosine kinases. It is an important regulator of various cellular processes including apoptosis, cell survival, cellular differentiation, cell migration, and transformation. One family of nonreceptor TKs includes the genes TEC (MIM 600583), TXK (MIM 600058), ITK (MIM 186973), and BTK (MIM 300300). All of these proteins are homologs of the Drosophila Src28 TK and contain an SH3 and SH2 domain upstream of the TK domain.[supplied by OMIM].
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P51813
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM tris HCl with 0.05% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 4 mM DTT, 50% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 660
Molecular Weight (g/mol) 83 kDa
Name Human BMX, His Tag
Quantity 10 μg
Regulatory Status For research use only. Not for use in diagnostic procedures
Gene Alias BMX; BMX non-receptor tyrosine kinase; bone marrow tyrosine kinase gene in chromosome x protein; Bone marrow tyrosine kinase gene in chromosome x protein homolog; BTK-like on x chromosome; cytoplasmic tyrosine-protein kinase BMX; epa3; Epithelial and endothelial tyrosine kinase; ETK; Etk/Bmx; Etk/Bmx cytosolic tyrosine kinase; NTK38; NTK38 tyrosine kinase; PSCTK2; PSCTK3; RGD1565643; Tyro8; TYRO8 protein tyrosine kinase 8
Common Name ETK
KinaseFamily TEC
Product Type BMX Recombinant Human Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System Baculovirus
Protein Family Kinase and inhibitors
Form Liquid
Protein Subtype Cytoplasmic tyrosine kinase
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.