Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BNC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575589
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575589 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575589 Supplier Invitrogen™ Supplier No. PA575589
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

Bnc 1 is a 994 amino acid transcription factor specific for squamous epithelium and for the constituent keratinocytes at a stage either prior to or at the very beginning of terminal differentiation. It is a zinc finger protein with three separated pairs of zinc fingers and a nuclear localization signal. Bnc 1 is a soluble protein that can shuttle between the nucleus and the cytoplasm, and its location depends on the proliferative potential of the cell. It is expressed relatively uniformly in the nucleoplasm, and phosphorylation on Ser-537 and Ser-541 leads to its cytoplasmic localization. It is present in the basal cell layer of the epidermis, in hair follicles and also in abundance in the germ cells of testis and ovary, and to a lower extent in thymus, spleen, mammary glands, placenta, brain and heart. Reports suggest that it plays a regulatory role in keratinocyte proliferation and also in rRNA transcription. It is also known to play a role in the differentiation of spermatozoa and oocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BNC1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene BNC1
Gene Accession No. O35914, Q01954
Gene Alias AI047752; AW546376; basonuclin 1; BNC; bnc 1; Bnc1; BSN1; HsT19447; zinc finger protein basonuclin-1
Gene Symbols BNC1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues in Human BNC1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12173, 365299, 646
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.