Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BNC2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584417
Description
Immunogen sequence: STQNEYNESS ESEVSPTPYK NDQTPNRNAL TSITNVEPKT EPACVSPIQN SAPVSDLTKT EHPKSSFRIH RMRRMGSASR KGRVFCNA.
Bnc2 gene ontology annotations related to this gene include nucleic acid binding.
Specifications
BNC2 | |
Polyclonal | |
Unconjugated | |
BNC2 | |
5031434M05Rik; 8430420F16Rik; basonuclin 2; bnc2; BNC2 (HGNC:30988); BSN2; zinc finger protein basonuclin-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54796 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6ZN30 | |
BNC2 | |
Recombinant protein corresponding to Human BNC2. Recombinant protein control fragment (Product #RP-108440). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction