Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BNC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113664
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113664 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113664 Supplier Invitrogen™ Supplier No. PA5113664
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GTLRVHYKTV HLREMHKCKV PGCNMMFSSV RSRNRHSQNP NLHKNIPFTS For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

Bnc2 gene ontology annotations related to this gene include nucleic acid binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BNC2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene BNC2
Gene Accession No. Q8BMQ3
Gene Alias 5031434M05Rik; 8430420F16Rik; basonuclin 2; bnc2; BNC2 (HGNC:30988); BSN2; zinc finger protein basonuclin-2
Gene Symbols BNC2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal region of Mouse Bnc2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 242509
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.