Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BNIP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP169182
Description
BNIP1 Polyclonal specifically detects BNIP1 in Human samples. It is validated for Western Blot.Specifications
| BNIP1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| BCL2/adenovirus E1B 19 kDa protein-interacting protein 1, BCL2/adenovirus E1B 19kDa interacting protein 1, BCL2/adenovirus E1B 19kD-interacting protein 1, NIP1, SEC20, SEC20L, Transformation-related gene 8 protein, TRG-8, vesicle transport protein SEC20 | |
| Rabbit | |
| 30 kDa | |
| 100 μL | |
| Apoptosis | |
| 662 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q12981-1 | |
| BNIP1 | |
| Synthetic peptides corresponding to BNIP1 (BCL2/adenovirus E1B 19kDa interacting protein 1) The peptide sequence was selected from the N terminal of BNIP1. Peptide sequence DFNSPTTPVTFSDLEQLAKEQDKESEKQLLLQEVENHKKQMLSNQASWRK. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Zebrafish: 75%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction