Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BNIP1 Monoclonal Antibody (OTI2E11), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA802193
Description
This gene is a member of the BCL2/adenovirus E1B 19 kd-interacting protein family. It interacts with the E1B 19 kDa protein, which protects cells from virally-induced cell death. The encoded protein also interacts with E1B 19 kDa-like sequences of BCL2, another apoptotic protector. In addition, this protein is involved in vesicle transport into the endoplasmic reticulum. Alternative splicing of this gene results in four protein products with identical N- and C-termini.Specifications
| BNIP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q12981 | |
| BNIP1 | |
| Human recombit protein fragment corresponding to amino acids 1-199 of human BNIP1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI2E11 | |
| Unconjugated | |
| BNIP1 | |
| NIP1, SEC20, TRG-8 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 662 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction