Learn More
BNIP1 Mouse anti-Human, Clone: OTI3G1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF802189

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene is a member of the BCL2/adenovirus E1B 19 kd-interacting protein family. It interacts with the E1B 19 kDa protein, which protects cells from virally-induced cell death. The encoded protein also interacts with E1B 19 kDa-like sequences of BCL2, another apoptotic protector. In addition, this protein is involved in vesicle transport into the endoplasmic reticulum. Alternative splicing of this gene results in four protein products with identical N- and C-termini.Specifications
BNIP1 | |
Monoclonal | |
Unconjugated | |
BNIP1 | |
NIP1, SEC20, TRG-8 | |
Mouse | |
Affinity Chromatography | |
RUO | |
662 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI3G1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q12981 | |
BNIP1 | |
Human recombit protein fragment corresponding to amino acids 1-199 of human BNIP1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.