Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BNIP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582335
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582335 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582335 Supplier Invitrogen™ Supplier No. PA582335
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KQMLSNQASW RKANLTCKIA IDNLEKAELL QGGDLLRQRK TTKESLAQTS STITESLMGI SRMMAQQVQQ SEEAMQSLVT SSRTILDANE EFKSMSGTIQ LGRKLITKYN RRELTDKL Highest antigen sequence indentity to the following orthologs: Mouse - 93%, Rat - 94%.

NIP1 (BNIP1) is a member of the BCL2/adenovirus E1B 19 kd-interacting protein (BNIP) family. It interacts with the E1B 19 kDa protein which is responsible for the protection of virally-induced cell death, as well as E1B 19 kDa-like sequences of BCL2, also an apoptotic protector. Alternative splicing of this gene results in four products of unknown function. Transcript variant BNIP1 contains the entire coding region of the gene. This variant contains a fully conserved BH3 domain, which has been associated with pro-apoptotic function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BNIP1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BNIP1
Gene Accession No. Q12981
Gene Alias 2010005M06Rik; 5930429G21Rik; BCL2 interacting protein 1; BCL2/adenovirus E1B 19 kDa protein-interacting protein 1; BCL2/adenovirus E1B 19kD interacting protein 1; BCL2/adenovirus E1B 19kDa interacting protein 1; BCL2/adenovirus E1B 19kDa-interacting protein 1; BCL2/adenovirus E1B 19kDa-interacting protein 1, NIP1; BCL2/adenovirus E1B interacting protein 1; BCL2/adenovirus E1B interacting protein 1, NIP1; BCL2/adenovirus E1B interacting protein 1-like 1; bnip1; bnip1 {ECO:0000312; Bnip1l1; EMBL:AAL50991.1, ECO:0000312; NIP1; RGD:620799}; Sec20; Sec20l; SNARE protein SEC20; transformation-related gene 8 protein; TRG8; TRG-8; Unknown (protein for MGC:133975); vesicle transport protein SEC20
Gene Symbols BNIP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BNIP1. Recombinant protein control fragment (Product #RP-104555).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 662
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.