Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BNIP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109246
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109246 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109246 Supplier Invitrogen™ Supplier No. PA5109246
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: Rat liver tissue lysate, HepG2, LOVO, human liver tissue, human placenta tissue.

NIP1 (BNIP1) is a member of the BCL2/adenovirus E1B 19 kd-interacting protein (BNIP) family. It interacts with the E1B 19 kDa protein which is responsible for the protection of virally-induced cell death, as well as E1B 19 kDa-like sequences of BCL2, also an apoptotic protector. Alternative splicing of this gene results in four products of unknown function. Transcript variant BNIP1 contains the entire coding region of the gene. This variant contains a fully conserved BH3 domain, which has been associated with pro-apoptotic function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BNIP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene BNIP1
Gene Accession No. Q12981
Gene Alias 2010005M06Rik; 5930429G21Rik; BCL2 interacting protein 1; BCL2/adenovirus E1B 19 kDa protein-interacting protein 1; BCL2/adenovirus E1B 19kD interacting protein 1; BCL2/adenovirus E1B 19kDa interacting protein 1; BCL2/adenovirus E1B 19kDa-interacting protein 1; BCL2/adenovirus E1B 19kDa-interacting protein 1, NIP1; BCL2/adenovirus E1B interacting protein 1; BCL2/adenovirus E1B interacting protein 1, NIP1; BCL2/adenovirus E1B interacting protein 1-like 1; bnip1; bnip1 {ECO:0000312; Bnip1l1; EMBL:AAL50991.1, ECO:0000312; NIP1; RGD:620799}; Sec20; Sec20l; SNARE protein SEC20; transformation-related gene 8 protein; TRG8; TRG-8; Unknown (protein for MGC:133975); vesicle transport protein SEC20
Gene Symbols BNIP1
Host Species Rabbit
Immunogen Recombinant protein within Human BN1P1 aa 27-228.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 662
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.