Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BNIP2 Antibody (OTI1C12), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270258UV
Description
BNIP2 Monoclonal specifically detects BNIP2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
BNIP2 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
BNIP2 | |
Full length recombinant protein of human BNIP2 (NP_004321) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot | |
OTI1C12 | |
Western Blot | |
BCL2/adenovirus E1B 19 kDa protein-interacting protein 2, BCL2/adenovirus E1B 19kDa interacting protein 2, BCL2/adenovirus E1B 19kD-interacting protein 2, BNIP-2, NIP2 | |
Mouse | |
Affinity Purified | |
Apoptosis | |
663.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction