Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BNIP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA592864
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA592864 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA592864 Supplier Invitrogen™ Supplier No. PA592864
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VELKEEWQDE DFPIPLPEDD SIEADILAIT GPEDQPGSLE VNGNKVRKKL MAPDISLTLD PSDGSVLSDD LDESGEIDLD GLDTPSENSN EFEWEDDLPK PKTTEVIRKG SITEYTAAEE KEDGRRWRMF RIGEQDHRVD MKAIEPYKKV ISHGGY; Positive Samples: U-937, SW480, NCI-H460, Mouse heart; Cellular Location: Cytoplasm, perinuclear region.

BNIP-2, (previously designated Nip2 and Nip21 in human and mouse respectively), shares homology with the non-catalytic domain of Cdc42 GTPase-activating protein (Cdc42GAP). Through binding to Cdc42GAP, BNIP-2 enhances the GTPase activity of Cdc42GAP, facilitating the hydrolysis of GTP bound to Cdc42 and thereby, mediating the signaling pathways involving receptor kinases, small GTPases and apoptotic proteins. Nix, which is also designated Nip3L or Bnip3L, is highly related to BNIP-3, and both proteins localize to the mitochondria where they associate with Bcl-2 proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BNIP2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.67 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Bnip2
Gene Accession No. O54940, Q12982
Gene Alias 5730523P12Rik; Bcl-2 and E1B 19 kDa interacting protein; BCL2 interacting protein 2; BCL2/adenovirus E1B 19 kDa protein-interacting protein 2; BCL2/adenovirus E1B 19 kDa-interacting protein 2 beta; BCL2/adenovirus E1B 19kDa interacting protein 2; BCL2/adenovirus E1B 19kDa-interacting protein 1, NIP2; BCL2/adenovirus E1B interacting protein 1, NIP2; BCL2/adenovirus E1B interacting protein 2; BNIP 2; BNIP2; BNIP-2; BNIP2beta; NIP2; Nip21; Nip2l
Gene Symbols Bnip2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 125-280 of human BNIP2 (NP_0043212).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12175, 663
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.