Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BNIP3 Monoclonal Antibody (ANa40)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA124688
Description
Positive Control: 293T cells transfected with BNIP3.
NIP3 is a member of the BCL2/adenovirus E1B 19 kd-interacting protein (BNIP) family. It interacts with the E1B 19 kDa protein which is responsible for the protection of virally-induced cell death, as well as E1B 19 kDa-like sequences of BCL2, also an apoptotic protector. NIP3 contains a BH3 domain and a transmembrane domain, which have been associated with pro-apoptotic function. The dimeric mitochondrial protein is known to induce apoptosis, even in the presence of BCL2.
Specifications
BNIP3 | |
Monoclonal | |
2.4 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
Q12983 | |
BNIP3 | |
Recombinant fragment corresponding to residues 1-163 of human BNIP3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry | |
ANa40 | |
Unconjugated | |
BNIP3 | |
BCL2 interacting protein 3; BCL2/adenovirus E1B 19 kDa protein-interacting protein 3; BCL2/adenovirus E1B 19 kDa-interacting protein 1, NIP3; BCL2/adenovirus E1B 19 kDa-interacting protein 3; BCL2/adenovirus E1B 19kDa interacting protein 3; BCL2/adenovirus E1B 19kDa-interacting protein 1, NIP3; BCL2/adenovirus E1B interacting protein 1, NIP3; BCL2/adenovirus E1B interacting protein 3; Bnip3; NIP3 | |
Mouse | |
Protein A | |
RUO | |
664 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction