Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BNIP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596388
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596388 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596388 Supplier Invitrogen™ Supplier No. PA596388
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AFPPPAAEAH PAARRGLRSP QLPSGAMSQN GAPGMQEESL QGSWVELHFS NNGNGGSVPA SVSIYNGDME KILLDAQHES G Positive Samples: A375, Mouse heart; Cellular Location: Mitochondrion, Mitochondrion outer membrane, Single-pass membrane protein.

NIP3 is a member of the BCL2/adenovirus E1B 19 kd-interacting protein (BNIP) family. It interacts with the E1B 19 kDa protein which is responsible for the protection of virally-induced cell death, as well as E1B 19 kDa-like sequences of BCL2, also an apoptotic protector. NIP3 contains a BH3 domain and a transmembrane domain, which have been associated with pro-apoptotic function. The dimeric mitochondrial protein is known to induce apoptosis, even in the presence of BCL2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BNIP3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene BNIP3
Gene Accession No. O55003, Q12983
Gene Alias BCL2 interacting protein 3; BCL2/adenovirus E1B 19 kDa protein-interacting protein 3; BCL2/adenovirus E1B 19 kDa-interacting protein 1, NIP3; BCL2/adenovirus E1B 19 kDa-interacting protein 3; BCL2/adenovirus E1B 19kDa interacting protein 3; BCL2/adenovirus E1B 19kDa-interacting protein 1, NIP3; BCL2/adenovirus E1B interacting protein 1, NIP3; BCL2/adenovirus E1B interacting protein 3; Bnip3; NIP3
Gene Symbols BNIP3
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12176, 664, 84480
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.