Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BNIP3 Rabbit anti-Human, Mouse, Rat, Clone: 7Q7V3, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | BNIP3 |
---|---|
Clone | 7Q7V3 |
Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
BNIP3 Monoclonal antibody specifically detects BNIP3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
BNIP3 | |
Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
BCL2/adenovirus E1B 19 kDa protein-interacting protein 3, BCL2/adenovirus E1B 19kDa interacting protein 3, BCL2/adenovirus E1B 19kD-interacting protein 3, NIP3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 40-120 of human BNIP3 (NP_004043.3). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
7Q7V3 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Apoptosis, Autophagy, Cancer, Hypoxia, Tumor Suppressors | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
664 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title