Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BNIP3L Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300002MFV610
Description
BNIP3L Polyclonal antibody specifically detects BNIP3L in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
BNIP3L | |
Polyclonal | |
Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry-Paraffin | |
Adenovirus E1B19K-binding protein B5, BCL2/adenovirus E1B 19 kDa protein-interacting protein 3A, BCL2/adenovirus E1B 19 kDa protein-interacting protein 3-like, BCL2/adenovirus E1B 19-kd protein-interacting protein 3a, BCL2/adenovirus E1B 19kDa interacting protein 3-like, BNIP3a, BNIP3H, NIP3L, NIP3-like protein X, Nix, NIXBCL2/adenovirus E1B 19kD-interacting protein 3-like | |
Produced in rabbits immunized with purified, recombinant Human BNIP3L (Accession#: NP_004322.1; Ser2-Lys187). | |
0.1 mL | |
Apoptosis, Autophagy, Cancer, Hypoxia, Mitophagy, p53 Pathway | |
665 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction