Learn More
Invitrogen™ BNIP3L Monoclonal Antibody (ZN002)

Description
BNIP3L Monoclonal Antibody for Western Blot, ICC/IF, ELISA
Specifications
Specifications
| Antigen | BNIP3L |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | ZN002 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.4 |
| Gene | BNIP3L |
| Gene Accession No. | O60238, Q9Z2F7 |
| Gene Alias | Adenovirus E1B19K-binding protein B5; BCL2 interacting protein 3 like; BCL2/adenovirus E1B 19 kDa protein-interacting protein 3A; BCL2/adenovirus E1B 19 kDa protein-interacting protein 3-like; BCL2/adenovirus E1B 19 kDa-interacting protein 3-like; BCL2/adenovirus E1B 19-kd protein-interacting protein 3a; BCL2/adenovirus E1B 19kDa interacting protein 3 like; BCL2/adenovirus E1B 19kDa interacting protein 3-like; BCL2/adenovirus E1B 19kDa-interacting protein 3-like; BCL2/adenovirus E1B 19kD-interacting protein 3-like; BCL2/adenovirus E1B interacting protein 3-like; BNIP3A; BNIP3H; BNIP3L; C86132; D14Ertd719e; Nip3L; NIP3-like protein X; NIX |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.