Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BNIPL Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$499.50
Specifications
| Antigen | BNIPL |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
BNIPL Polyclonal specifically detects BNIPL in Human samples. It is validated for Western Blot.Specifications
| BNIPL | |
| Polyclonal | |
| Rabbit | |
| Cytokine Research | |
| bcl-2/adenovirus E1B 19 kDa-interacting protein 2-like protein, BCL2/adenovirus E1B 19kD interacting protein like, BNIP-2 similar, BNIPl-1, BNIPL-2, BNIP-S, PP753 | |
| BNIPL | |
| IgG | |
| 40 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| NP_612122 | |
| 149428 | |
| Synthetic peptide directed towards the middle region of human BNIPLThe immunogen for this antibody is BNIPL. Peptide sequence AENYLLVHLSGGTSRAQVPPLSWIRQCYRTLDRRLRKNLRALVVVHATWY. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title