Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BNIPL Monoclonal Antibody (2C8)
GREENER_CHOICE

Catalog No. PIMA524526
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524526 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524526 Supplier Invitrogen™ Supplier No. MA524526
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MRKRLSAPEL RLSLTKGPGN DGASPTQSAP SSPDGSSDLE IDELETPSDS EQLDSGHEFE WEDELPRAEG LGTSETAERL GRGCMWDVTG EDGHHWRVFR MGPREQRVDM TVIEPYKKVL SHGGYHGDGL NAVILFASCY LPRSSIPNYT YVMEHLFRYM VGTLELLVAE NYLLVHLSGG TSRAQVPPLS WIRQCYRTLD RRLRKNLRAL VVVHATWYVK AFLALLRPFI SSKFTRKIRF LDSLGELAQL ISLDQVHIPE AVRQLDRDLH GSGGT.

May be a bridge molecule between BCL2 and ARHGAP1/CDC42 in promoting cell death.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BNIPL
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2C8
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene BNIPL
Gene Accession No. Q7Z465
Gene Alias BCL2 interacting protein like; Bcl-2/adenovirus E1B 19 kDa-interacting protein 2-like protein; BCL2/adenovirus E1B 19kD interacting protein like; BNIP-2 similar; BNIPL; BNIPL1; BNIPL-1; BNIPL2; BNIPL-2; BNIPS; BNIP-S; BNIP-Salpha; BNIP-Sbeta; PP753
Gene Symbols BNIPL
Host Species Mouse
Immunogen BNIPL (AAH27868, 1 a.a. ~ 275 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 149428
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.