Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BNIPL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103648
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103648 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103648 Supplier Invitrogen™ Supplier No. PA5103648
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total BNIPL.

May be a bridge molecule between BCL2 and ARHGAP1/CDC42 in promoting cell death.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BNIPL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene BNIPL
Gene Accession No. Q7Z465
Gene Alias BCL2 interacting protein like; Bcl-2/adenovirus E1B 19 kDa-interacting protein 2-like protein; BCL2/adenovirus E1B 19kD interacting protein like; BNIP-2 similar; BNIPL; BNIPL1; BNIPL-1; BNIPL2; BNIPL-2; BNIPS; BNIP-S; BNIP-Salpha; BNIP-Sbeta; PP753
Gene Symbols BNIPL
Host Species Rabbit
Immunogen A synthesized peptide derived from human BNIPL(Accession Q7Z465), corresponding to amino acid residues R81-D131.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 149428
Target Species Human, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.