Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BNP Antibody (50E1cc), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB200440
Description
BNP Monoclonal specifically detects BNP in Human samples. It is validated for Western Blot, ELISA, Immunoassay, Sandwich ELISA.Specifications
BNP | |
Monoclonal | |
Unconjugated | |
P16860 | |
NPPB | |
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with synthetic human BNP, whole molecule, conjugated with carrier protein. | |
Protein A purified | |
RUO | |
Primary | |
Detects Human BNP and proBNP | |
Store at 4C. Do not freeze. | |
IgG1 |
Western Blot, Flow Cytometry, ELISA, Immunoassay, CyTOF | |
500 | |
Western Blot 1:100-1:2000, Flow Cytometry, ELISA 1:100-1:2000, Immunoassay, Sandwich ELISA 1:100-1:2000, CyTOF-ready | |
BNP, brain type natriuretic peptide, Gamma-brain natriuretic peptide, natriuretic peptide B, natriuretic peptide precursor B, natriuretic peptides B, natriuretic protein | |
Mouse | |
14.7 kDa | |
0.2 mg | |
Cytokine Research, Immunology, Neuroscience, Neurotransmission | |
4879 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction