Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BNP Antibody (B1865M) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB6001183
Description
BNP Monoclonal specifically detects BNP in Human samples. It is validated for Western Blot, ELISA.Specifications
BNP | |
Monoclonal | |
7.0 mg/mL | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
NPPB | |
Synthetic BNP FGRKMDRISSSS, conjugated with carrier protein. Hybridization of Sp2/0 myeloma cells with spleen cells from BALB/c mice | |
0.2 mg | |
Primary | |
Human BNP and proBNP | |
Store at 4C. Do not freeze. | |
IgG1 |
Western Blot, ELISA | |
24C5 | |
Unconjugated | |
BNP, brain type natriuretic peptide, Gamma-brain natriuretic peptide, natriuretic peptide B, natriuretic peptide precursor B, natriuretic peptides B, natriuretic protein | |
Mouse | |
Immunogen affinity purified | |
RUO | |
4879 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction