Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BOB1 Antibody (OTI1E11), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271223AF405
Description
BOB1 Monoclonal specifically detects BOB1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
BOB1 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
POU2AF1 | |
Full length recombinant protein of human BOB1 (NP_006226) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot | |
OTI1E11 | |
Western Blot | |
B-cell-specific coactivator OBF-1, BOB-1, OBF-1, OBF1BOB1, OCAB, OCA-B, OCT-binding factor 1, POU class 2 associating factor 1, POU domain class 2, associating factor 1, POU domain class 2-associating factor 1, POU domain, class 2, associating factor 1 | |
Mouse | |
Affinity Purified | |
Immunology | |
5450.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction