Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BOC-beta-alanine, 99%

Catalog No. p-4228406
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle

CAS: 3303-84-2 | C8H15NO4 | 189.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3303-84-2
Molecular Formula C8H15NO4
Molecular Weight (g/mol) 189.21
MDL Number MFCD00037291
InChI Key WCFJUSRQHZPVKY-UHFFFAOYSA-N
Synonym boc-beta-alanine, boc-beta-ala-oh, n-boc-beta-alanine, boc-b-ala-oh, boc-beta-ala, n-tert-butoxycarbonyl-beta-alanine, 3-tert-butoxycarbonyl amino propanoic acid, n-t-boc-beta-alanine, n-t-butyloxycarbonyl-beta-alanine, 3-tert-butoxycarbonylamino propanoic acid
PubChem CID 76809
IUPAC Name 3-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NCCC(O)=O

Specifications

Melting Point 77.5°C to 78.5°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Quantity 5 g
Formula Weight 189.21
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material BOC-β-Alanine
EINECSNumber 221-979-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.