Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+/-)-Boc-alpha-phosphonoglycine trimethyl ester, 95%

Catalog No. AAB2542106
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 89524-98-1 | C10H20NO7P | 297.24 g/mol

It is used as Wittig-Horner reagent for preparing (Z)-Boc-protected dehydroamino acid derivatives and saturated, unnatural amino acids by subsequent asymmetric hydrogenation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as Wittig-Horner reagent for preparing (Z)-Boc-protected dehydroamino acid derivatives and saturated, unnatural amino acids by subsequent asymmetric hydrogenation.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89524-98-1
Molecular Formula C10H20NO7P
Molecular Weight (g/mol) 297.24
MDL Number MFCD02178893
InChI Key LJHAPRKTPAREGO-ZETCQYMHSA-N
Synonym methyl 2-tert-butoxycarbonyl amino-2-dimethoxyphosphoryl acetate, +/--boc-alpha-phosphonoglycine trimethyl ester, n-boc-2-phosphonoglycine trimethyl ester, methyl n-boc-2-dimethylphosphono glycinate, methyl 2-tert-butoxycarbonylamino-2-dimethoxyphosphoryl-acetate, boc-alpha-phosphonoglycine trimethyl ester, n-butoxycarbonyl trimethylphosphonoglycine, methyl 2-tert-butoxy carbonyl amino-2-dimethoxyphosphoryl acetate, boc-a-phosphonoglycine trimethyl ester, boc-?-phosphonoglycine trimethyl ester
PubChem CID 397380
IUPAC Name methyl 2-dimethoxyphosphoryl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetate
SMILES COC(=O)[C@@H](NC(=O)OC(C)(C)C)P(=O)(OC)OC

Specifications

Melting Point 67°C to 71°C
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 297.24
Percent Purity 95%
Chemical Name or Material (+/-)-Boc-alpha-phosphonoglycine trimethyl ester
Hazard Category H301
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.