Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Boc-D(OMe)-FMK Inhibitor
SDP

Applications: In vitro assay, In vivo assay, Block/Neutralize

$402.00 - $1049.00

Specifications

Color White
For Use With (Application) Blocking/Neutralizing, In vitro assay, In vivo assay
Content And Storage Store at -20°C. Avoid freeze-thaw cycles
Product Type Boc-D(OMe)-FMK Inhibitor
Molecular Weight (g/mol) 263
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP2293955
SDP
View Documents
Novus Biologicals™
NBP2293955MG
5 mg
Each for $1,049.00
Only null left
Add to Cart
 
NBP229395
SDP
View Documents
Novus Biologicals™
NBP229395
1 mg
Each for $402.00
Only null left
Add to Cart
 
Description

Description

Specifications

Specifications

White
Store at -20°C. Avoid freeze-thaw cycles
263
Semi-solid
Blocking/Neutralizing, In vitro assay, In vivo assay
Boc-D(OMe)-FMK Inhibitor
Boc-D(OMe)-FMK
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.