Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Boc-D(OMe)-FMK Inhibitor
SDP

Catalog No. NBP2293955 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2293955 5 mg
NBP229395 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP2293955 Supplier R&D Systems™ Supplier No. NBP2293955MG

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Applications: In vitro assay, In vivo assay, Block/Neutralize

Specifications

Color White
For Use With (Application) Blocking/Neutralizing, In vitro assay, In vivo assay
Content And Storage Store at -20°C. Avoid freeze-thaw cycles
Quantity 5 mg
Product Type Boc-D(OMe)-FMK Inhibitor
Molecular Weight (g/mol) 263
Product Line Novus
Inhibitors Boc-D(OMe)-FMK
Form Semi-solid

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.