Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BOC-D-alpha-phenylglycine, 99%

Catalog No. p-4228423
Click to view available options
Quantity:
1 g
5 g

CAS: 33125-05-2 | C13H17NO4 | 251.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33125-05-2
Molecular Formula C13H17NO4
Molecular Weight (g/mol) 251.28
MDL Number MFCD00062043,MFCD00065588
InChI Key HOBFSNNENNQQIU-UHFFFAOYNA-N
Synonym boc-d-phg-oh, boc-d-phenylglycine, n-boc-d-phenylglycine, n-boc-d-2-phenylglycine, n-boc-l-phenylglycine, n-tert-butoxycarbonyl-d-2-phenylglycine, boc-d-alpha-phenylglycine, r-tert-butoxycarbonyl amino phenyl acetic acid, r-tert-butoxycarbonylamino-phenyl-acetic acid, r-2-tert-butoxycarbonyl amino-2-phenylacetic acid
PubChem CID 2755953
SMILES CC(C)(C)OC(=O)NC(C(O)=O)C1=CC=CC=C1

Specifications

Melting Point 88°C to 92°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass Bottle
Specific Rotation -145
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-2-phenylacetic acid
Quantity 1 g
Formula Weight 251.28
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material BOC-D-α-phenylglycine
Show More Show Less
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.