Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BOC-L-Phenylalanine, 99+%

Catalog No. p-3776621
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle

CAS: 13734-34-4 | C14H19NO4 | 265.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13734-34-4
Molecular Formula C14H19NO4
Molecular Weight (g/mol) 265.31
MDL Number MFCD00002663
InChI Key ZYJPUMXJBDHSIF-NSHDSACASA-N
Synonym boc-phe-oh, boc-l-phenylalanine, boc-phenylalanine, n-tert-butoxycarbonyl-l-phenylalanine, n-boc-l-phenylalanine, n-t-boc-l-phenylalanine, n-alpha-t-boc-l-phenylalanine, n-t-butyloxycarbonyl-l-phenylalanine, l-phenylalanine, n-1,1-dimethylethoxy carbonyl, boc-phe
PubChem CID 77857
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 85.0°C to 88.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass Bottle
Linear Formula C6H5CH2CH(COOH)NHCOOC(CH3)3
Specific Rotation + 25.00
Beilstein 14, 1595
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol and methylene chloride, insoluble in aliphatic hydrocarbons
Water 0.5% max. (K.F.)
IUPAC Name (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-phenylpropanoic acid
Quantity 5 g
Formula Weight 265.3
Percent Purity ≥99%
Physical Form Fine Crystalline Powder
Chemical Name or Material BOC-L-Phenylalanine
Show More Show Less
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.