Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sebacoyl chloride, 92%, technical
Click to view available options
Quantity:
50 g
250 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -2.5°C |
| Color | White to Beige |
| Density | 1.1210g/mL |
| Boiling Point | 220.0°C (75.0 mmHg) |
| Flash Point | >110°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 90% min. (GC) |
| Packaging | Glass bottle |
| Refractive Index | 1.4670 to 1.4700 |
| Linear Formula | ClCO(CH2)8COCl |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Harmful if swallowed. Fatal in contact with skin. Causes severe skin burns and eye damage. Contact with water liberates toxic gas. May be corrosive to metals. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breath |
| EINECSNumber | 203-843-4 |
| RTECSNumber | HD8454250 |
| TSCA | TSCA |
| Recommended Storage | Product may develop some turbidity or precipitate |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction