Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BOC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547308
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547308 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547308 Supplier Invitrogen™ Supplier No. PA547308
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In Western blots, approximately 25% cross-reactivity with recombinant human BOC is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

CDON and BOC are cell surface receptors of the immunoglobulin repeat family involved in myogenic differentiation. CDON and BOC are coexpressed during development, form complexes with each other in a cis fashion, and are related to each other in their ectodomains, but each has a unique long cytoplasmic tail.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BOC
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Boc
Gene Accession No. Q6AZB0
Gene Alias 4732455C11; biregional Cdon binding protein; biregional cell adhesion molecule-related/down-regulated by oncogenes (Cdon) binding protein; BOC; BOC cell adhesion associated, oncogene regulated; Boc homolog; Boc homolog (mouse); brother of CDO; brother of CDON; CDON2; cell adhesion associated, oncogene regulated 2; mFLJ00376; Protein BOC; UNQ604/PRO1190
Gene Symbols Boc
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse BOC Asp26-Lys508 (Gln ins) Val509-Asp846.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117606
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.