Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BOC Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109452
Description
Immunogen sequence: MLRGTMTAWR GMRPEVTLAC LLLATAGCFA DLNEVPQVTV QPASTVQKPG GTVILGCVVE PPRMNVTWRL NGKELNGSDD ALGVLITHGT LVITALNNHT VGRYQCVARM PAGAVASVPA TVTLA.
CDON and BOC are cell surface receptors of the immunoglobulin repeat family involved in myogenic differentiation. CDON and BOC are coexpressed during development, form complexes with each other in a cis fashion, and are related to each other in their ectodomains, but each has a unique long cytoplasmic tail.
Specifications
| BOC | |
| Polyclonal | |
| Unconjugated | |
| Boc | |
| 4732455C11; biregional Cdon binding protein; biregional cell adhesion molecule-related/down-regulated by oncogenes (Cdon) binding protein; BOC; BOC cell adhesion associated, oncogene regulated; Boc homolog; Boc homolog (mouse); brother of CDO; brother of CDON; CDON2; cell adhesion associated, oncogene regulated 2; mFLJ00376; Protein BOC; UNQ604/PRO1190 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 117606, 360715, 91653 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.45 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6AZB0, Q9BWV1 | |
| Boc | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-125 of human BOC (NP_150279.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction