Learn More
Boekel Scientific™ Open Source Bio™ (OSB) Transmitter with pH Probe
SDPSCI_ED
Catalog No. s21151 Shop All Boekel Scientific Products
Click to view available options
Range (pH):
0.001 to 14.000
Catalog No. S21151
Supplier: Boekel Scientific™ 555001/EMD
Includes
Open Source Transmitter, pH probe, calibration reagents and temperature probe.
A flexible device for measuring pH.
Open Source Bio or OSB probes and transmitters work just like conventional probes and transmitters but allow for additional functionality because the equipment is built around an Arduino™ compatible board. The transmitter is capable of single parameter measurement of pH, DO and Conductivity. The transmitter includes high precision temperature probe for temperature compensation.
  • Built around UNO, an open source circuit board that allows easy user programming and experiment control
  • Repairable and industrial design supports years of experiments
  • High precision temperature probe for temperature compensation
  • External I/O connector for connection to pumps or alarms
  • User friendly probe calibration menus
  • USB drive data logging
  • Large color screen
Specifications:
  • Range: 0.001 to 14.000
  • Accuracy: ±0.02
  • Calibration: 3-Point
  • Also works with DO and EC probes
Warranty and Services
Two years.
Specifications
Accuracy (pH) ±0.02
Calibration Points (pH) 3
Display Type Large Screen
Range (pH) 0.001 to 14.000
Includes Open Source Transmitter, pH probe, calibration reagents and temperature probe.
Product Type OSB Transmitter with pH supplies and Temperature probe
Warranty 2 Years
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.