Learn More
Innovating Science™ Boiling Chips
Boiling chips for laboratory use.
$4.54 - $7.04
Specifications
Type | Boiling Chips |
---|---|
Material | Borosilicate Glass |
Diameter (Metric) | 9.5 mm |
For Use With (Equipment) | Chemware Glassware |
Alert:
Warning: May cause damage to organs (lungs) through prolonged or repeated exposure (inhalation). WARNING! This product can expose you to a chemical, crystalline silica, which is known to the State of California to cause cancer.
Description
Boiling chips for laboratory use.
Specifications
Boiling Chips | |
Borosilicate Glass | |
9.5 mm | |
Chemware Glassware |