Description
These inert non-volatile granules resist most chemical action except those of concentrated sulfuric acids and strong alkalies. And, they won't contaminate distillate.
This product is for educational use - no C of A is available.
Specifications
Specifications
| Type | Boiling Chips |
| Material | Marble |
| Quantity | 100 g |