Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BOK Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA126427
Description
PA1-26427 detects Bok from rat samples.
The protein encoded by this gene belongs to the BCL-2 protein family. BCL-2 family members form hetero- or homodimers and act as anti- and pro-apoptotic regulators that are involved in a wide variety of cellular activities. This protein contains all four BCL-2 like domains and is a pro-apoptotic BCL-2 protein identified in the ovary.
Specifications
BOK | |
Polyclonal | |
Unconjugated | |
Bok | |
AI847676; apoptosis activator Mtd; AU021146; BCL2 family apoptosis regulator BOK; BCL2 related ovarian killer; BCL2L9; bcl2-L-9; Bcl-2-like protein 9; BCL2-related ovarian killer; bcl-2-related ovarian killer protein; BCL2-related ovarian killer protein; BOK; BOK, BCL2 family apoptosis regulator; Bok-BH3; BOKL; hBOK; matador; mtd; Protein matador | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29884, 51800 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.2 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide, 0.02% Thimerosal; pH 7.2 | |
O35425, Q792S6 | |
Bok | |
Synthetic peptide surrounding amino acid 44 of rat Bok. | |
50 μg | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction