Learn More
Invitrogen™ BOK Polyclonal Antibody

Description
The antibody detects endogenous levels of total BOK protein.
Specifications
Specifications
| Antigen | BOK |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | Bok |
| Gene Accession No. | Q9UMX3 |
| Gene Alias | AI847676; apoptosis activator Mtd; AU021146; BCL2 family apoptosis regulator BOK; BCL2 related ovarian killer; BCL2L9; bcl2-L-9; Bcl-2-like protein 9; BCL2-related ovarian killer; bcl-2-related ovarian killer protein; BCL2-related ovarian killer protein; BOK; BOK, BCL2 family apoptosis regulator; Bok-BH3; BOKL; hBOK; matador; mtd; Protein matador |
| Gene Symbols | Bok |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.