Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BOK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142612
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142612 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142612 Supplier Invitrogen™ Supplier No. PA5142612
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:8,000.

The protein encoded by this gene belongs to the BCL-2 protein family. BCL-2 family members form hetero- or homodimers and act as anti- and pro-apoptotic regulators that are involved in a wide variety of cellular activities. This protein contains all four BCL-2 like domains and is a pro-apoptotic BCL-2 protein identified in the ovary.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BOK
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Bok
Gene Accession No. Q9UMX3
Gene Alias AI847676; apoptosis activator Mtd; AU021146; BCL2 family apoptosis regulator BOK; BCL2 related ovarian killer; BCL2L9; bcl2-L-9; Bcl-2-like protein 9; BCL2-related ovarian killer; bcl-2-related ovarian killer protein; BCL2-related ovarian killer protein; BOK; BOK, BCL2 family apoptosis regulator; Bok-BH3; BOKL; hBOK; matador; mtd; Protein matador
Gene Symbols Bok
Host Species Goat
Immunogen Peptide with sequence C-HISLQSEPVVTD (aa 93-104).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 666
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.