Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BOK Rabbit anti-Human, Mouse, Rat, Clone: S03-1B6, Novus Biologicals™

Rabbit Monoclonal Antibody
$263.00 - $573.00
Specifications
Antigen | BOK |
---|---|
Clone | S03-1B6 |
Dilution | Western Blot, ELISA, Immunocytochemistry/Immunofluorescence 2.5 μg/mL |
Applications | Western Blot, ELISA, Immunofluorescence |
Classification | Monoclonal |
Description
BOK Monoclonal antibody specifically detects BOK in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
BOK | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence 2.5 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
Bcl2-L-9, BCL2L9bcl-2-related ovarian killer protein, Bcl-2-like protein 9, BCL2-related ovarian killer, BOKL, hBOK, MGC4631 | |
A synthetic peptide of human BOK (Uniprot#: Q9UMX3) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
S03-1B6 | |
Western Blot, ELISA, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Apoptosis | |
50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
666 | |
IgG | |
Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title