Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BOLA1 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299968JF646
Description
BOLA1 Polyclonal antibody specifically detects BOLA1 in Human samples. It is validated for ELISASpecifications
BOLA1 | |
Polyclonal | |
ELISA | |
bolA homolog 1 (E. coli), bolA-like 1, bolA-like 1 (E. coli), bolA-like protein 1, CGI-143, hBolA, MGC75015, RP11-196G18.18 | |
Produced in rabbits immunized with purified, recombinant Human BOLA1 (Uniprot#: Q9Y3E2; Met1-Pro137). | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
51027 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction